Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,6-Dichloro-3-fluoro-5-(trifluoromethyl)pyridine

Catalog No. MNO0001731G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MNO0001731G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MNO0001731G Supplier Sigma Aldrich Supplier No. MNO0001731G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl cis-3-iodoacrylate (cis-ethyl-α-iodoacrylate, ethyl (z)-α-iodoacrylate) is z-vinyl iodide derivative. it is reported to be prepared from ethyl propiolate by treating with sodium iodide. its suzuki coupling reaction with cyclic vinyl boronic acids has been studied.ethyl cis-3-iodoacrylate may be used in the synthesis of the following:
  • Vinyl sulfides
  • (z)-1-iodohept-1-en-3-ol
  • (e)-1-iodohept-1-en-3-ol
  • (z)-α-iodoacrolein
  • 6-oxo-m1guo

Specifications

Molecular Formula C6HCl2F4N
Linear Formula C6HCl2F4N
MDL Number MFCD11100554
Quantity 5 g
Molecular Weight (g/mol) 233.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.