Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-3-methylaniline, 99+%, Thermo Scientific™

Catalog No. AC206480100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC206480100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC206480100 Supplier Thermo Scientific Chemicals Supplier No. 206480100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64063-37-2
Molecular Formula C7H7Cl2N
Molecular Weight (g/mol) 176.04
MDL Number MFCD00007676
InChI Key AYVZXDFCFQSWJD-UHFFFAOYSA-N
Synonym benzenamine, 2,6-dichloro-3-methyl, 2, 6-dichloro-3-methylaniline, 2,6-dichloro-3-methylphenylamine, 2,6-dichloro-m-toluidine, unii-t7qd52545o, 2,6-dichlor-3-methylanilin, 2,6-dichloro-3-methylbenzenamine, acmc-1b8ol, dsstox_cid_24710, dsstox_rid_80412
PubChem CID 116529
IUPAC Name 2,6-dichloro-3-methylaniline
SMILES CC1=CC=C(Cl)C(N)=C1Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 37°C to 40°C
Color Beige-Brown to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Linear Formula Cl2C6H2(CH3)NH2
Quantity 10 g
Formula Weight 176.04
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dichloro-3-methylaniline, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.