Learn More
Medchemexpress LLC 2,6-Dichloro-3-pyridinamine | 62476-56-6 | 163.01 | 1 ML
SDP
Catalog No. 5000447156
Encompass_Preferred
2,6-Dichloro-3-pyridinamine is a biochemical reagent designed for life science-related research. This organic compound is intended for research use only and is not for patient use. It appears as a solid with a brown to reddish-brown color.
  • Used as a biological material
  • Suitable for life science research
  • Solid appearance
  • Brown to reddish-brown color
  • Store at 4°C, protected from light (solid)
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light
Catalog No. 50-004-47156 Supplier Medchemexpress LLC Supplier No. HYW00213610MM/1ML
2,6-Dichloro-3-pyridinamine is a biochemical reagent designed for life science-related research. This organic compound is intended for research use only and is not for patient use. It appears as a solid with a brown to reddish-brown color.
  • Used as a biological material
  • Suitable for life science research
  • Solid appearance
  • Brown to reddish-brown color
  • Store at 4°C, protected from light (solid)
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light