Learn More
Medchemexpress LLC 2,6-Dichloro-3-pyridinamine | 62476-56-6 | 163.01 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00213610G
Encompass_Preferred
2,6-Dichloro-3-pyridinamine is a biochemical reagent suitable for life science related research. This organic compound is intended for research use only.
  • Can be used as a biological material.
  • Appears as a solid with a brown to reddish brown color.
  • Store at 4°C, protected from light.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Suitable for biochemical assay reagents.
Catalog No. 50-004-47361