Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,6-dichloro-4-[2-(1-piperazinyl)-4-pyridinyl]-N-(1,3,5-trimethyl-1H-pyrazol-4-yl)-benzenesulfonam | 1215010-55-1 | 99.7% | 495.43 | C21H24Cl2N6O2S | 100 MG
SDP

Catalog No. 5000246015
Encompass_Preferred

DDD85646 is a small-molecule N-myristoyltransferase (NMT) inhibitor supplied for research use. It shows low-nanomolar activity against Trypanosoma brucei NMT and is used in biochemical, cellular, and in vivo studies when formulated per supplier guidance.

  • Potent NMT inhibitor with reported low-nanomolar potency.
  • Solid, light yellow to green yellow powder; molecular weight 495.43.
  • High purity (≈99.7%) suitable for research applications.
  • Soluble in DMSO at 100 mg/mL; in vivo formulations reported using PEG300, Tween-80, or SBE-β-CD.
  • Storage recommendations: powder -20 °C (long term) or 4 °C (short term); stock solutions at -80 °C or -20 °C.

Catalog No. 50-002-46015 Supplier Medchemexpress LLC Supplier No. HY103056100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DDD85646 is a small-molecule N-myristoyltransferase (NMT) inhibitor supplied for research use. It shows low-nanomolar activity against Trypanosoma brucei NMT and is used in biochemical, cellular, and in vivo studies when formulated per supplier guidance.

  • Potent NMT inhibitor with reported low-nanomolar potency.
  • Solid, light yellow to green yellow powder; molecular weight 495.43.
  • High purity (≈99.7%) suitable for research applications.
  • Soluble in DMSO at 100 mg/mL; in vivo formulations reported using PEG300, Tween-80, or SBE-β-CD.
  • Storage recommendations: powder -20 °C (long term) or 4 °C (short term); stock solutions at -80 °C or -20 °C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.