Learn More
Medchemexpress LLC 2,6-dichloro-4-[2-(1-piperazinyl)-4-pyridinyl]-N-(trimethylpyrazol-4-yl)-benzenesulfonamide | 1215010-55-1 | 99.7% | 495.43 g/mol | C21H24Cl2N6O2S | 25 MG
SDP
Catalog No. 5000246455
Encompass_Preferred
DDD85646 (IMP-366) is an orally active inhibitor of N-myristoyltransferase (NMT) with potent enzymatic activity against Trypanosoma brucei (TbNMT IC50 = 2 nM) and measurable activity versus human NMT (hNMT IC50 = 4 nM). It is supplied as a high-purity research compound for in vitro and in vivo pharmacology studies targeting NMT.
  • Potent N-myristoyltransferase inhibition (TbNMT IC50 = 2 nM; hNMT IC50 = 4 nM).
  • High purity suitable for research (reported ≥99.7%).
  • Soluble in DMSO (100 mg/mL); recommended in vivo formulations provided for dosing.
  • Available in small research pack sizes, suitable for preclinical studies.
  • Powder stable at -20°C for long-term storage; solutions should be aliquoted and stored cold.
Catalog No. 50-002-46455 Supplier Medchemexpress LLC Supplier No. HY10305625MG
DDD85646 (IMP-366) is an orally active inhibitor of N-myristoyltransferase (NMT) with potent enzymatic activity against Trypanosoma brucei (TbNMT IC50 = 2 nM) and measurable activity versus human NMT (hNMT IC50 = 4 nM). It is supplied as a high-purity research compound for in vitro and in vivo pharmacology studies targeting NMT.
  • Potent N-myristoyltransferase inhibition (TbNMT IC50 = 2 nM; hNMT IC50 = 4 nM).
  • High purity suitable for research (reported ≥99.7%).
  • Soluble in DMSO (100 mg/mL); recommended in vivo formulations provided for dosing.
  • Available in small research pack sizes, suitable for preclinical studies.
  • Powder stable at -20°C for long-term storage; solutions should be aliquoted and stored cold.