Learn More
Medchemexpress LLC 2,6-dichloro-4-(2-piperazin-1-ylpyridin-4-yl)-n-(1,3,5-trimethyl-1h-pyrazol-4-yl)benzenesulfonamide | 1215010-55-1 | 99.7% | 495.43 g/mol | C21H24Cl2N6O2S | 5 MG
SDP
Catalog No. 5000246186
Encompass_Preferred
2,6-dichloro-4-(2-piperazin-1-ylpyridin-4-yl)-N-(1,3,5-trimethyl-1H-pyrazol-4-yl)benzenesulfonamide is a small-molecule N-myristoyltransferase (NMT) inhibitor with potent enzymatic and cellular activity against Trypanosoma brucei. It demonstrates nanomolar potency and is supplied in solid and DMSO solution formulations for research use.
  • Potent NMT inhibition; TbNMT IC50 = 2 nM, hNMT IC50 = 4 nM.
  • Orally active in preclinical models.
  • High purity (≈99.7%) suitable for biochemical assays.
  • Available as solids and DMSO solutions for assay flexibility.
  • Useful for parasite biology and antitrypanosomal drug discovery research.
Catalog No. 50-002-46186 Supplier Medchemexpress LLC Supplier No. HY1030565MG
2,6-dichloro-4-(2-piperazin-1-ylpyridin-4-yl)-N-(1,3,5-trimethyl-1H-pyrazol-4-yl)benzenesulfonamide is a small-molecule N-myristoyltransferase (NMT) inhibitor with potent enzymatic and cellular activity against Trypanosoma brucei. It demonstrates nanomolar potency and is supplied in solid and DMSO solution formulations for research use.
  • Potent NMT inhibition; TbNMT IC50 = 2 nM, hNMT IC50 = 4 nM.
  • Orally active in preclinical models.
  • High purity (≈99.7%) suitable for biochemical assays.
  • Available as solids and DMSO solutions for assay flexibility.
  • Useful for parasite biology and antitrypanosomal drug discovery research.