Learn More
Medchemexpress LLC 2,6-dichloro-4-(2-piperazin-1-ylpyridin-4-yl)-N-(1,3,5-trimethylpyrazol-4-yl)benzenesulfonamide | 1215010-55-1 | 99.7% | 495.43 g·mol⁻¹ | C21H24Cl2N6O2S | 50 MG
SDP
Catalog No. 5000247409
Encompass_Preferred
  • Potent N-myristoyltransferase inhibitor, IC50 ≈ 2 nM against TbNMT.
  • Low-nanomolar activity against human NMT for comparative studies.
  • High purity (~99.7%) suitable for biochemical and in vivo research.
  • Solid powder with light yellow to green-yellow appearance.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (2 years).
  • Available in multiple quantities, including 50 MG for small-scale studies.
Catalog No. 50-002-47409 Supplier Medchemexpress LLC Supplier No. HY10305650MG
  • Potent N-myristoyltransferase inhibitor, IC50 ≈ 2 nM against TbNMT.
  • Low-nanomolar activity against human NMT for comparative studies.
  • High purity (~99.7%) suitable for biochemical and in vivo research.
  • Solid powder with light yellow to green-yellow appearance.
  • Stable when stored as powder at -20°C (up to 3 years) or 4°C (2 years).
  • Available in multiple quantities, including 50 MG for small-scale studies.