Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-4-(trifluoromethyl)pyridin-3-amine, 97%, Thermo Scientific™

Catalog No. SPB06360EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SPB06360EA 10 g Amber Glass Bottle
SPB06360DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SPB06360EA Supplier Thermo Scientific Chemicals Supplier No. SPB06360EA
Add to Cart

Chemical Identifiers

CAS 175277-67-5
Molecular Formula C6H3Cl2F3N2
Molecular Weight (g/mol) 231.00
MDL Number MFCD00173957
InChI Key BVJJNBRZQVLPIC-UHFFFAOYSA-N
PubChem CID 2735898
IUPAC Name 2,6-dichloro-4-(trifluoromethyl)pyridin-3-amine
SMILES NC1=C(Cl)N=C(Cl)C=C1C(F)(F)F

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 2,6-dichloro-4-trifluoromethyl pyridin-3-amine, 3-amino-2,6-dichloro-4-trifluoromethyl pyridine, 3-amino-2,6-dichloro-4-trifluoromethyl-pyridine, pubchem2205, 2,6-dichloro-3-amino-4-trifluoromethylpyridine, 2,6-dichloro-4-trifluoromethyl-3-pyridinamine, 2,6-dichloro-4-trifluoromethyl-3-pyridylamine, 2,6-dichloro-4-trifluoromethyl-pyridin-3-ylamine, 3-pyridinamine,2,6-dichloro-4-trifluoromethyl, 3-pyridinamine, 2,6-dichloro-4-trifluoromethyl
Formula Weight 231
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloro-4-(trifluoromethyl)pyridin-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.