Learn More
TARGETMOL CHEMICALS INC 2 6-Dichloro-N-2-cyclopropanec
SDP
Catalog No. 502877539
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 2,6-Dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide (GDC046), a potent lead analog, has good kinase selectivity, physicochemical properties and pharmacokinetic profile. Purity 99.65%
Catalog No. 50-287-7539 Supplier TARGETMOL CHEMICALS INC Supplier No. T223385MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 2,6-Dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide (GDC046), a potent lead analog, has good kinase selectivity, physicochemical properties and pharmacokinetic profile. Purity 99.65%