Learn More
Medchemexpress LLC 2,6-dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide | 1258292-64-6 | 98.1% | 10 MG
SDP
Catalog No. 5000232561
Encompass_Preferred
GDC-046 (2,6-dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide) is a small-molecule, orally bioavailable TYK2 inhibitor used for preclinical kinase research, supplied as a solid for laboratory studies.
  • Potent TYK2 inhibition with low-nanomolar Ki values (TYK2 4.8 nM; JAK1 0.7 nM; JAK2 0.7 nM; JAK3 0.4 nM).
  • Suitable for biochemical and cellular kinase assays.
  • Solid form, commonly provided in 10 mg quantities.
  • High purity reported (~98.1%).
  • Molecular formula C16H13Cl2N3O2 and molecular weight 350.2 g/mol.
Catalog No. 50-002-32561 Supplier Medchemexpress LLC Supplier No. HY12046910MG
GDC-046 (2,6-dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide) is a small-molecule, orally bioavailable TYK2 inhibitor used for preclinical kinase research, supplied as a solid for laboratory studies.
  • Potent TYK2 inhibition with low-nanomolar Ki values (TYK2 4.8 nM; JAK1 0.7 nM; JAK2 0.7 nM; JAK3 0.4 nM).
  • Suitable for biochemical and cellular kinase assays.
  • Solid form, commonly provided in 10 mg quantities.
  • High purity reported (~98.1%).
  • Molecular formula C16H13Cl2N3O2 and molecular weight 350.2 g/mol.