Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,6-dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide | 1258292-64-6 | 99.8% | 1 ML
SDP

Catalog No. 5000234320
Encompass_Preferred

GDC-046 (2,6-dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide) is a potent, selective, orally bioavailable TYK2 inhibitor with reported Ki values of 4.8 nM (TYK2), 0.7 nM (JAK1), 0.7 nM (JAK2), and 0.4 nM (JAK3). Supplied for research use, available as a 10 mM solution in DMSO (1 mL) or as solid material.

  • Potent, selective TYK2 inhibitor with low-nanomolar activity against JAK family kinases.
  • Molecular weight 350.2 g/mol; formula C16H13Cl2N3O2.
  • CAS 1258292-64-6; purity 99.8%.
  • Physical form solid, off-white to yellow; also supplied as 10 mM solution in DMSO (1 mL).
  • Solubility in DMSO 62.5 mg/mL (178.47 mM); ultrasonic assistance recommended.
  • Storage: solid at 4 °C; in solvent store at -80 °C (up to 6 months) or -20 °C (up to 1 month); protect from light.

Catalog No. 50-002-34320 Supplier Medchemexpress LLC Supplier No. HY12046910MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GDC-046 (2,6-dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide) is a potent, selective, orally bioavailable TYK2 inhibitor with reported Ki values of 4.8 nM (TYK2), 0.7 nM (JAK1), 0.7 nM (JAK2), and 0.4 nM (JAK3). Supplied for research use, available as a 10 mM solution in DMSO (1 mL) or as solid material.

  • Potent, selective TYK2 inhibitor with low-nanomolar activity against JAK family kinases.
  • Molecular weight 350.2 g/mol; formula C16H13Cl2N3O2.
  • CAS 1258292-64-6; purity 99.8%.
  • Physical form solid, off-white to yellow; also supplied as 10 mM solution in DMSO (1 mL).
  • Solubility in DMSO 62.5 mg/mL (178.47 mM); ultrasonic assistance recommended.
  • Storage: solid at 4 °C; in solvent store at -80 °C (up to 6 months) or -20 °C (up to 1 month); protect from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.