Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzaldehyde, 97+%

Catalog No. AAA1329536
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1329536 500 g
AAA1329514 25 g
AAA1329522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1329536 Supplier Thermo Scientific Chemicals Supplier No. A1329536
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 83-38-5 | C7H4Cl2O | 175.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83-38-5
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003307
InChI Key DMIYKWPEFRFTPY-UHFFFAOYSA-N
Synonym benzaldehyde, 2,6-dichloro, ccris 6014, 2,6-dichloro benzaldehyde, 2,6-dichloro-benzaldehyde, 2,6-dichloro-benzaldehyd, 2,6-dichlorbenzaldehyd, pubchem8225, acmc-1bkey, 2,6dichloro-benzaldehyde, benzaldehyde,6-dichloro
PubChem CID 6737
IUPAC Name 2,6-dichlorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Cl)C=O)Cl

Specifications

Melting Point 69°C to 73°C
Boiling Point 124°C (10 mmHg)
Flash Point 135°C (275°F)
Quantity 500 g
UN Number UN3261
Beilstein 386477
Sensitivity Air Sensitive
Formula Weight 175.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorobenzaldehyde
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 201-472-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.