Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzoxazole 98.0+%, TCI America™
SDP

Catalog No. D345925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D345925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D345925G Supplier TCI America Supplier No. D345925G

missing translation for 'validMainframeMsg'

Specifications

CAS 3621-82-7
Melting Point 50°C
Color White-Yellow
Boiling Point 110°C
Molecular Formula C7H3Cl2NO
MDL Number MFCD07368635
Quantity 25 g
InChI Key LVVQTPZQNHQLOM-UHFFFAOYSA-N
SMILES C1=CC2=C(C=C1Cl)OC(=N2)Cl
IUPAC Name 2,6-dichloro-1,3-benzoxazole
Molecular Weight (g/mol) 188.007
PubChem CID 77175
Formula Weight 188.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorobenzoxazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.