Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. D174725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D174725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D174725G Supplier TCI America Supplier No. D174725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15258-73-8
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00004609
InChI Key WKKHCCZLKYKUDN-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzyl alcohol, 2,6-dichlorophenyl methanol, benzenemethanol, 2,6-dichloro, 2,6-dichlorobenzenemethanol, benzyl alcohol, 2,6-dichloro, 2,6-dcal, 2,6-dichlorobenzylalcohol, benzenemethanol, 2,6-dichloro-9ci, 2,6-dichloro-benzenemethanol, 2,6-dichloro-phenyl-methanol
PubChem CID 27156
IUPAC Name (2,6-dichlorophenyl)methanol
SMILES C1=CC(=C(C(=C1)Cl)CO)Cl

Specifications

Melting Point 98°C
Color White
Quantity 25 g
Formula Weight 177.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorobenzyl Alcohol
RTECSNumber DO0900000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.