Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzyl Bromide 98.0+%, TCI America™
SDP

Catalog No. D174925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D174925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D174925G Supplier TCI America Supplier No. D174925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20443-98-5
Molecular Formula C7H5BrCl2
Molecular Weight (g/mol) 239.92
MDL Number MFCD00000577
InChI Key PDFGFQUSSYSWNI-UHFFFAOYSA-N
Synonym 2,6-dichlorobenzyl bromide, 2-bromomethyl-1,3-dichlorobenzene, alpha-bromo-2,6-dichlorotoluene, 2,6-dichlorobenzylbromide, benzene, 2-bromomethyl-1,3-dichloro, 1-bromomethyl-2,6-dichlorobenzene, .alpha.-bromo-2,6-dichlorotoluene, a-bromo-2,6-dichlorotoluene, 2,6 dichloro benzyl bromide, benzene, 2-bromomethyl-1,3-dichloro-9ci
PubChem CID 30159
IUPAC Name 2-(bromomethyl)-1,3-dichlorobenzene
SMILES ClC1=CC=CC(Cl)=C1CBr

Specifications

Melting Point 57°C
Color White
Boiling Point 135°C
Quantity 25 g
UN Number 3261
Formula Weight 239.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorobenzyl Bromide
RTECSNumber XS7966000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.