Learn More
Medchemexpress LLC 2,6-Dichlorodiphenylamine | 15307-93-4 | 99.78% | 238.11 | 100 MG
SDP
Catalog No. 5000453652
Encompass_Preferred
2,6-Dichlorodiphenylamine is an analogue of Diclofenac Sodium and exhibits anti-Candida albicans activity. Diclofenac Sodium is described as a potent and nonselective anti-inflammatory agent that acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells.
  • Analogue of Diclofenac Sodium.
  • Exhibits anti-Candida albicans activity.
  • Potent and nonselective anti-inflammatory agent.
  • Acts as a COX inhibitor.
Catalog No. 50-004-53652 Supplier Medchemexpress LLC Supplier No. HYW012126100MG
2,6-Dichlorodiphenylamine is an analogue of Diclofenac Sodium and exhibits anti-Candida albicans activity. Diclofenac Sodium is described as a potent and nonselective anti-inflammatory agent that acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells.
  • Analogue of Diclofenac Sodium.
  • Exhibits anti-Candida albicans activity.
  • Potent and nonselective anti-inflammatory agent.
  • Acts as a COX inhibitor.