Learn More
Medchemexpress LLC 2,6-Dichlorodiphenylamine | 15307-93-4 | 99.8% | 238.12 | 1 ML
SDP
Catalog No. 5000452420
Encompass_Preferred
2,6-Dichlorodiphenylamine is an analogue of Diclofenac Sodium and demonstrates anti-Candida albicans activity. Diclofenac Sodium is a potent and nonselective anti-inflammatory agent, functioning as a COX inhibitor with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, respectively. This product is intended for research use only and is not sold to patients.
  • Analogue of diclofenac sodium
  • Demonstrates anti-Candida albicans activity
  • Potent and nonselective anti-inflammatory agent
  • Functions as a COX inhibitor
  • High purity of 99.78%
  • Causes skin irritation
  • Causes serious eye irritation
  • Very toxic to aquatic life with long lasting effects
  • Store at 4°C, protected from light
  • Avoid inhalation, contact with eyes and skin
  • Avoid dust and aerosol formation
  • Use in areas with appropriate exhaust ventilation
Catalog No. 50-004-52420 Supplier Medchemexpress LLC Supplier No. HYW01212610MM/1ML
2,6-Dichlorodiphenylamine is an analogue of Diclofenac Sodium and demonstrates anti-Candida albicans activity. Diclofenac Sodium is a potent and nonselective anti-inflammatory agent, functioning as a COX inhibitor with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, respectively. This product is intended for research use only and is not sold to patients.
  • Analogue of diclofenac sodium
  • Demonstrates anti-Candida albicans activity
  • Potent and nonselective anti-inflammatory agent
  • Functions as a COX inhibitor
  • High purity of 99.78%
  • Causes skin irritation
  • Causes serious eye irritation
  • Very toxic to aquatic life with long lasting effects
  • Store at 4°C, protected from light
  • Avoid inhalation, contact with eyes and skin
  • Avoid dust and aerosol formation
  • Use in areas with appropriate exhaust ventilation