Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorofluorobenzene 98.0+%, TCI America™
SDP

Catalog No. D24855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24855G 5 g
D248525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D24855G Supplier TCI America Supplier No. D24855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2268-05-5
Molecular Formula C6H3Cl2F
Molecular Weight (g/mol) 164.99
MDL Number MFCD00010307
InChI Key JORVCRLRRRRLFI-UHFFFAOYSA-N
Synonym 2,6-dichlorofluorobenzene, 1,3-dichloro-2-fluoro-benzene, 2,6-dichloro-1-fluorobenzene, 2,6-dichloro fluoro benzene, benzene,1,3-dichloro-2-fluoro, benzene, 1,3-dichloro-2-fluoro, pubchem2311, 2,6-dichlorfluorbenzol, acmc-2097ae, 2,6-dichloro fluorobenzene
PubChem CID 137520
IUPAC Name 1,3-dichloro-2-fluorobenzene
SMILES C1=CC(=C(C(=C1)Cl)F)Cl

Specifications

Melting Point 39°C
Boiling Point 171°C
Quantity 5 g
UN Number 1325
Formula Weight 164.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorofluorobenzene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.