Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorophenyl isocyanate, 98%

Catalog No. AAL1042106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1042106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1042106 Supplier Thermo Scientific Chemicals Supplier No. L1042106
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 39920-37-1 | C7H3Cl2NO | 188.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39920-37-1
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.007
MDL Number MFCD00002003
InChI Key HMVKMAMIRAVXAN-UHFFFAOYSA-N
Synonym 2,6-dichlorophenyl isocyanate, benzene, 1,3-dichloro-2-isocyanato, 2,6-dichlorophenylisocyanate, isocyanic acid 2,6-dichlorophenyl ester, pubchem23916, acmc-1ailv, 2,6 dichlorophenylisocyanate, 2,6-dichlorobenzenisocyanate, 2,6dichlorophenyl isocyanate, 1,3-dichloro-2-isocyanato-benzene
PubChem CID 604537
IUPAC Name 1,3-dichloro-2-isocyanatobenzene
SMILES C1=CC(=C(C(=C1)Cl)N=C=O)Cl

Specifications

Melting Point 42°C to 46°C
Boiling Point 101°C (5 mmHg)
Flash Point 76°C (168°F)
Quantity 5 g
UN Number UN2250
Beilstein 608323
Sensitivity Moisture sensitive
Formula Weight 188.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichlorophenyl isocyanate
Hazard Category H301+H311+H331-H315-H319-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROPHENYL ISOCYANATES
EINECSNumber 254-699-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.