Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorophenylacetone 98.0+%, TCI America™
SDP

Catalog No. D29295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29295G Supplier TCI America Supplier No. D29295G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 93457-06-8
Molecular Formula C9H8Cl2O
Molecular Weight (g/mol) 203.062
MDL Number MFCD03410452
InChI Key YAEXJQJVTXDTJM-UHFFFAOYSA-N
Synonym 2,6-dichlorophenylacetone, 1-2,6-dichlorophenyl propan-2-one, 1-2,6-dichlorophenyl acetone, acmc-209rlv, ghl.pd_mitscher_leg0.494, 1-2,6-dichlorophenyl-2-propanone
PubChem CID 2734103
IUPAC Name 1-(2,6-dichlorophenyl)propan-2-one
SMILES CC(=O)CC1=C(C=CC=C1Cl)Cl

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 105°C
Quantity 5 g
Formula Weight 203.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichlorophenylacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.