Learn More
Cayman Chemical 2 6-DIchloropurIn-9D-rIbo 25mg
SDP
Catalog No. 502990953
Encompass_Preferred
A building block; has been used in the synthesis of photoaffinity probes for nucleotide binding sites in proteins
Catalog No. 50-299-0953 Supplier Cayman Chemical Supplier No. 3140525 MG
A building block; has been used in the synthesis of photoaffinity probes for nucleotide binding sites in proteins