Learn More
Cayman Chemical 2 6-DIchloropurIn-9D-rIbo 50mg
SDP
Catalog No. 502990698
Encompass_Preferred
A building block; has been used in the synthesis of photoaffinity probes for nucleotide binding sites in proteins
Catalog No. 50-299-0698 Supplier Cayman Chemical Supplier No. 3140550 MG
A building block; has been used in the synthesis of photoaffinity probes for nucleotide binding sites in proteins