Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,6-dichloropyridine-3-boronic acid

Catalog No. CDS007852 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS007852 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS007852 Supplier Sigma Aldrich Supplier No. CDS007852250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl 2-bromo-2-butenoate(methyl (z)-2-bromocrotonate) yields (s)-2-bromobutanoic acid by bakerαs yeast fermentation
  • It undergoes biotransformation catalyzed by old yellow enzyme to yield methyl (s)-2-bromobutanoate, a key intermediate for the synthesis of chiral drugs
  • It is a biannelating reagent which participates in double michael-additions

Specifications

CAS 148493-34-9
Molecular Formula C5H4BCl2NO2
Linear Formula C5H4BCl2NO2
MDL Number MFCD03094989
Quantity 5 mL
Molecular Weight (g/mol) 191.81
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.