Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloropyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193911 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47071G 1 g
D4707-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47071G Supplier TCI America Supplier No. D47071G
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material 2,6-Dichloropyridine-3-boronic Acid (contains varying amounts of Anhydride)
CAS 148493-34-9
Melting Point 150°C
Color White-Yellow
Molecular Formula C5H4BCl2NO2
Quantity 1 g
MDL Number MFCD03094989
Synonym 2,6-dichloropyridine-3-boronic acid, 2,6-dichloropyridin-3-yl boronic acid, 2,6-dichloro-3-pyridineboronic acid, 3-borono-2,6-dichloropyridine, 2,6-dichloropyridin-3-yl-3-boronic acid, 2,6-dichloro-3-pyridinyl-boronic acid, boronic acid, 2,6-dichloro-3-pyridinyl, 2,6-dichloropyridine-3-boronicacid, pubchem5039, acmc-209d0p
InChI Key XBBLBQZAVMHEER-UHFFFAOYSA-N
SMILES B(C1=C(N=C(C=C1)Cl)Cl)(O)O
IUPAC Name (2,6-dichloropyridin-3-yl)boronic acid
Molecular Weight (g/mol) 191.802
PubChem CID 2762712
Formula Weight 191.80
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.