Learn More
Accela Chembio Inc 2 | 6-dichloropyridine-3-carbonitrile | 5g | 40381-90-6 | MFCD03411341 | 97+% | Shelf Life: 1440 Days | Light Sensitive
Supplier:  Accela Chembio Inc SY0065135G
Encompass_Preferred
2 | 6-dichloropyridine-3-carbonitrile | 5g | 40381-90-6 | MFCD03411341 | 97+% | Shelf Life: 1440 Days | Light Sensitive
Catalog No. 50-354-528