Learn More
Medchemexpress LLC 2 6-Dichloroquinone- 250mg
SDP
Catalog No. 5000204601
Encompass_Preferred
2 6-Dichloroquinone-4-chloroimide is a spray reagent for organic compounds 2 6-Dichloroquinone-4-chloroimide can be used in thin-layer chromatograms and detecting phenol and its derivatives 2 6-Dichloroquinone-4-chloroimide can be used as an optical sensor for rapid detection of permethrin in treated wood[1][2][3][4]
Catalog No. 50-002-04601 Supplier Medchemexpress LLC Supplier No. 250MGHYW105135
2 6-Dichloroquinone-4-chloroimide is a spray reagent for organic compounds 2 6-Dichloroquinone-4-chloroimide can be used in thin-layer chromatograms and detecting phenol and its derivatives 2 6-Dichloroquinone-4-chloroimide can be used as an optical sensor for rapid detection of permethrin in treated wood[1][2][3][4]