Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diethylaniline, 99.7%, For GC analysis, MP Biomedicals™

Catalog No. ICN15088625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15088625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15088625 Supplier MP Biomedicals, Inc Supplier No. 0215088625
Add to Cart
Add to Cart

Chemical Identifiers

CAS 579-66-8
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
InChI Key FOYHNROGBXVLLX-UHFFFAOYSA-N
Synonym benzenamine, 2,6-diethyl, 2,6-diethylbenzenamine, aniline, 2,6-diethyl, 2,6-diethyl aniline, 2-amino-1,3-diethylbenzene, 2,6-diethyl-aniline, ccris 2688, benzamine, 2,6-diethyl, 2,6 diethylaniline, 2,6-diethylphenylamine
PubChem CID 11369
ChEBI CHEBI:27880
IUPAC Name 2,6-diethylaniline
SMILES CCC1=C(C(=CC=C1)CC)N

Specifications

Color Yellow
Quantity 25 g
Formula Weight 149.2
Physical Form Liquid
Chemical Name or Material 2, 6-Diethylaniline, For GC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.