Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorobenzyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. D25075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25075G 5 g
D250725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25075G Supplier TCI America Supplier No. D25075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19064-18-7
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00004603
InChI Key LVICICZQETYOGS-UHFFFAOYSA-N
Synonym 2,6-difluorobenzyl alcohol, 2,6-difluorophenyl methanol, benzenemethanol, 2,6-difluoro, 2,6-difluorobenzylalcohol, 2,6-difluorobenzenemethanol, 2,6-difluoro-phenyl-methanol, 2,6-difluoro benzyl alcohol, 2,6-difluorophenyl methan-1-ol, pubchem4914, acmc-209etz
PubChem CID 87921
IUPAC Name (2,6-difluorophenyl)methanol
SMILES C1=CC(=C(C(=C1)F)CO)F

Specifications

Color Yellow
Boiling Point 88°C
Quantity 5 g
Formula Weight 144.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Difluorobenzyl Alcohol
EINECSNumber (3)-4498
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.