Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorobenzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D23375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23375G 5 g
D233725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23375G Supplier TCI America Supplier No. D23375G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 697-73-4
Molecular Formula C7H5ClF2
Molecular Weight (g/mol) 162.564
MDL Number MFCD00191346
InChI Key MJXRENZUAQXZGJ-UHFFFAOYSA-N
PubChem CID 581570
IUPAC Name 2-(chloromethyl)-1,3-difluorobenzene
SMILES C1=CC(=C(C(=C1)F)CCl)F

Specifications

Quantity 5 g
UN Number 3261
Synonym 2,6-difluorobenzyl chloride, 2-chloromethyl-1,3-difluorobenzene, 2,6-difluorobenzylchloride, benzene, 2-chloromethyl-1,3-difluoro, alpha-chloro-2,6-difluorotoluene, 2, 6-difluorobenzyl chloride, 1-chloromethyl-2,6-difluorobenzene, 2-chlormethyl-1,3-difluorbenzol, pubchem2178
Formula Weight 162.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Difluorobenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.