Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorophenol, 98%

Catalog No. p-3757699
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC311630050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC311630050 Supplier Thermo Scientific Chemicals Supplier No. 311630050
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 28177-48-2 | C6H4F2O | 130.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28177-48-2
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.1
MDL Number MFCD00002158
InChI Key CKKOVFGIBXCEIJ-UHFFFAOYSA-N
Synonym phenol, 2,6-difluoro, 2.6-difluorophenol, 2,6-difluoro-phenol, 2,6-difluorophenol, 2,6-difluoro phenol, pubchem1495, ksc493c1n, ckkovfgibxceij-uhfffaoysa
PubChem CID 94392
IUPAC Name 2,6-difluorophenol
SMILES C1=CC(=C(C(=C1)F)O)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38°C to 42°C
Color Colorless
Boiling Point 59°C to 61°C (17.0 mmHg)
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula F2C6H3OH
Quantity 5 g
Formula Weight 130.1
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2, 6-Difluorophenol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Flammable solid.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 248-884-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.