Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,6-Difluoropyridine

Catalog No. 2332775G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-327-75G 5 g
23-327-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-327-75G Supplier Sigma Aldrich Supplier No. 2332775G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Lithium diisopropylamide (lda)-mediated ortholithiations of 2,6-difluoropyridine in tetrahydrofuran at -78c has been studied using a combination of IR and NMR spectroscopic and computational methods
  • Polycondensation of bistrimethylsilyl derivatives of various diphenols with 2,6-difluoropyridine in n-methylpyrrolidone in the presence of k2co3 has been investigated.2,6-difluoropyridine has been used in the preparation of poly(pyridine ether)s via polycondensation with silylated 1,1,1-tris(4-hydroxyphenyl)ethane

Specifications

CAS 1513-65-1
Density 1.268 g/mL (at 25°C (literature))
Boiling Point 124.5°C (743 mmHg)
Molecular Formula C5H3F2N
Refractive Index n20/D 1.437 (literature)
Linear Formula C5H3F2N
MDL Number MFCD00006227
Quantity 5 g
Molecular Weight (g/mol) 115.08
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.