Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dihydroxyanthracene 95.0+%, TCI America™
SDP

Catalog No. D5151200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5151200MG 200 mg
D51511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5151200MG Supplier TCI America Supplier No. D5151200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 101488-73-7
Molecular Formula C14H10O2
Molecular Weight (g/mol) 210.232
InChI Key JBBHFHMVEOHFRE-UHFFFAOYSA-N
Synonym 2,6-Anthracenediol
PubChem CID 13466096
IUPAC Name anthracene-2,6-diol
SMILES C1=CC(=CC2=CC3=C(C=C21)C=C(C=C3)O)O

Specifications

Melting Point 300°C
Quantity 200 mg
Formula Weight 210.23
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dihydroxyanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.