Learn More
Selleck Chemical LLC 2.6-Dihydroxyanthraquinone S5169-25mg
SDP
Catalog No. 503108808
Encompass_Preferred
2 6-Dihydroxyanthraquinone (Anthraflavic acid Anthraflavin) is a potent and specific inhibitor of cytochrome P-448 activity
Catalog No. 50-310-8808 Supplier Selleck Chemical LLC Supplier No. S516925MG
2 6-Dihydroxyanthraquinone (Anthraflavic acid Anthraflavin) is a potent and specific inhibitor of cytochrome P-448 activity