Learn More
Selleck Chemical LLC 2 6-Dihydroxybenzoic acid
SDP
Catalog No. 5000003802
Encompass_Preferred
2 6-Dihydroxybenzoic acid ( -resorcylic acid 2-Carboxyresorcinol 2 6-Resorcylic acid) is a phenolic compound which is known to have poor biological performance such as DPPH scavenging activity and microbial growth inhibition
Catalog No. 50-000-03802 Supplier Selleck Chemical LLC Supplier No. S603125MG
2 6-Dihydroxybenzoic acid ( -resorcylic acid 2-Carboxyresorcinol 2 6-Resorcylic acid) is a phenolic compound which is known to have poor biological performance such as DPPH scavenging activity and microbial growth inhibition