Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dihydroxybenzoic acid, 97%

Catalog No. AC114881000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC114881000 100 g Plastic bottle
AC114880250 25 g Plastic bottle
AC114885000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC114881000 Supplier Thermo Scientific Chemicals Supplier No. 114881000
Add to Cart

CAS: 303-07-1 | C7H6O4 | 154.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-07-1
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00002462
InChI Key AKEUNCKRJATALU-UHFFFAOYSA-N
Synonym gamma-resorcylic acid, 2-carboxyresorcinol, 2,6-resorcylic acid, 6-hydroxysalicylic acid, benzoic acid, 2,6-dihydroxy, .gamma.-resorcylic acid, unii-rsa5g6vrpv, rsa5g6vrpv, 2,6-dihydroxy benzoic acid, 2,6-dihydroxy-benzoic acid
PubChem CID 9338
ChEBI CHEBI:68465
IUPAC Name 2,6-dihydroxybenzoic acid
SMILES OC(=O)C1=C(O)C=CC=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 160°C to 165°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula (HO)2C6H3CO2H
Quantity 100 g
Beilstein 10, 388
Solubility Information Solubility in water: 25g/L (25°C). Other solubilities: soluble in ethanol and ether
Formula Weight 154.12
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dihydroxybenzoic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-134-8
RTECSNumber DG8578000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.