Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dihydroxyquinoline 98.0+%, TCI America™
SDP

Catalog No. D28225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28225G 5 g
D28221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28225G Supplier TCI America Supplier No. D28225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19315-93-6
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00239446
InChI Key AQLYZDRHNHZHIS-UHFFFAOYSA-N
Synonym 6-Hydroxy-2(1H)-quinolinone, 2,6-Quinolinediol
PubChem CID 177065
ChEBI CHEBI:48996
IUPAC Name 6-hydroxy-1H-quinolin-2-one
SMILES C1=CC2=C(C=CC(=O)N2)C=C1O

Specifications

Quantity 5 g
Formula Weight 161.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dihydroxyquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.