Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diiodo-4-nitroaniline 98.0+%, TCI America™
SDP

Catalog No. D47365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47365G 5 g
D473625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47365G Supplier TCI America Supplier No. D47365G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5398-27-6
Molecular Formula C6H4I2N2O2
Molecular Weight (g/mol) 389.919
MDL Number MFCD00007683
InChI Key YPVYMWQYENWFAT-UHFFFAOYSA-N
PubChem CID 79386
IUPAC Name 2,6-diiodo-4-nitroaniline
SMILES C1=C(C=C(C(=C1I)N)I)[N+](=O)[O-]

Specifications

Melting Point 255°C
Quantity 5 g
Formula Weight 389.92
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diiodo-4-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.