Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diisopropylnaphthalene 99.0+%, TCI America™
SDP

Catalog No. D159825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D159825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D159825G Supplier TCI America Supplier No. D159825G

missing translation for 'validMainframeMsg'

Specifications

CAS 24157-81-1
Melting Point 70°C
Color White-Yellow
Molecular Formula C16H20
Quantity 25 g
MDL Number MFCD00021648
Synonym 2,6-diisopropylnaphthalene, naphthalene, 2,6-bis 1-methylethyl, 2,6-bis 1-methylethyl naphthalene, 2,6-di-iso-propylnaphthalene, 2,6-dipn, naphthalene, 2,6-diisopropyl, unii-1x71yeu9qb, 2,6-di propan-2-yl naphthalene, epa pesticide chemical code 055803, 1x71yeu9qb
InChI Key GWLLTEXUIOFAFE-UHFFFAOYSA-N
SMILES CC(C)C1=CC2=C(C=C1)C=C(C=C2)C(C)C
IUPAC Name 2,6-di(propan-2-yl)naphthalene
Molecular Weight (g/mol) 212.336
PubChem CID 32241
Formula Weight 212.34
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diisopropylnaphthalene
Show More Show Less
EINECSNumber (4)-0961
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.