Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxy-1,4-benzoquinone 97.0+%, TCI America™
SDP

Catalog No. D27065G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27065G Supplier TCI America Supplier No. D27065G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 530-55-2
Color Yellow
Molecular Formula C8H8O4
Quantity 5 g
MDL Number MFCD00016368
Synonym 2,6-dimethoxy-1,4-benzoquinone, 2,6-dimethoxy-p-benzoquinone, 2,6-dimethoxyquinone, 2,6-dimethoxybenzoquinone, 2,6-dimethoxy-p-quinone, 2,6-dimethoxysemiquinone anions, 2,5-cyclohexadiene-1,4-dione, 2,6-dimethoxy, dmbq, p-benzoquinone, 2,6-dimethoxy, 2,6-dimethoxybenzo-1,4-quinone
InChI Key OLBNOBQOQZRLMP-UHFFFAOYSA-N
SMILES COC1=CC(=O)C=C(C1=O)OC
IUPAC Name 2,6-dimethoxycyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol) 168.148
PubChem CID 68262
ChEBI CHEBI:27651
Formula Weight 168.15
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethoxy-1,4-benzoquinone
Show More Show Less
RTECSNumber DK4750000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.