Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxy-4-methylphenol 97.0+%, TCI America™
SDP

Catalog No. D259125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D259125G 25 g
D25915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D259125G Supplier TCI America Supplier No. D259125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6638-05-7
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00017289
InChI Key ZFBNNSOJNZBLLS-UHFFFAOYSA-N
Synonym 4-methyl-2,6-dimethoxyphenol, 4-methylsyringol, 2,6-dimethoxy-p-cresol, phenol, 2,6-dimethoxy-4-methyl, p-cresol, 2,6-dimethoxy, 3,5-dimethoxy-4-hydroxytoluene, 4-hydroxy-3,5-dimethoxytoluene, unii-fjj5mye75w, fjj5mye75w, 2,6-dimethoxy-4-methyl-phenol
PubChem CID 240925
IUPAC Name 2,6-dimethoxy-4-methylphenol
SMILES CC1=CC(=C(C(=C1)OC)O)OC

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 150°C
Quantity 25 g
Formula Weight 168.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,6-Dimethoxy-4-methylphenol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.