Learn More
Medchemexpress LLC 2,6-Dimethoxyphenol | 91-10-1 | 154.16 | 10 G
SDP
Catalog No. 5000449113
Encompass_Preferred
2,6-Dimethoxyphenol (Syringol) is a phenolic compound known for its ability to scavenge free radicals and exhibit antioxidant activity. It serves as a substrate for determining laccase activity and has been shown to inhibit the formation of N-nitrosomorpholine (NMOR) in the stomach and blood of Sprague-Dawley rats.
  • Scavenges free radicals
  • Exhibits antioxidant activity
  • Substrate for determining laccase activity
  • Inhibits N-nitrosomorpholine (NMOR) formation in rat models
Catalog No. 50-004-49113 Supplier Medchemexpress LLC Supplier No. HYW00397210G
2,6-Dimethoxyphenol (Syringol) is a phenolic compound known for its ability to scavenge free radicals and exhibit antioxidant activity. It serves as a substrate for determining laccase activity and has been shown to inhibit the formation of N-nitrosomorpholine (NMOR) in the stomach and blood of Sprague-Dawley rats.
  • Scavenges free radicals
  • Exhibits antioxidant activity
  • Substrate for determining laccase activity
  • Inhibits N-nitrosomorpholine (NMOR) formation in rat models