Learn More
Medchemexpress LLC 2,6-Dimethoxyphenol | 91-10-1 | 154.16 | 500 MG
SDP
Catalog No. 5000446514
Encompass_Preferred
2,6-Dimethoxyphenol (Syringol) is a phenolic compound known for its ability to scavenge free radicals and exhibit antioxidant activity. It serves as a valuable reagent in various biochemical applications, particularly as a substrate for determining laccase activity.
  • Phenolic compound
  • Scavenges free radicals
  • Exhibits antioxidant activity
  • Used as a substrate for determining laccase activity
Catalog No. 50-004-46514 Supplier Medchemexpress LLC Supplier No. HYW003972500MG
2,6-Dimethoxyphenol (Syringol) is a phenolic compound known for its ability to scavenge free radicals and exhibit antioxidant activity. It serves as a valuable reagent in various biochemical applications, particularly as a substrate for determining laccase activity.
  • Phenolic compound
  • Scavenges free radicals
  • Exhibits antioxidant activity
  • Used as a substrate for determining laccase activity