Learn More
Medchemexpress LLC 2,6-Dimethoxyphenol (Syringol) | 91-10-1 | 154.16 | 5 G
SDP
Catalog No. 5000445729
Encompass_Preferred
2,6-Dimethoxyphenol (Syringol) is a phenolic compound that scavenges free radicals and exhibits antioxidant activity. It is also a substrate for determining laccase activity.
  • Scavenges free radicals and exhibits antioxidant activity.
  • Functions as a substrate for determining laccase activity.
  • Solid appearance.
  • Off-white to light yellow color.
  • Ships at room temperature (continental US).
  • For research use only.
Catalog No. 50-004-45729 Supplier Medchemexpress LLC Supplier No. HYW0039725G
2,6-Dimethoxyphenol (Syringol) is a phenolic compound that scavenges free radicals and exhibits antioxidant activity. It is also a substrate for determining laccase activity.
  • Scavenges free radicals and exhibits antioxidant activity.
  • Functions as a substrate for determining laccase activity.
  • Solid appearance.
  • Off-white to light yellow color.
  • Ships at room temperature (continental US).
  • For research use only.