Learn More
Medchemexpress LLC 2,6-Dimethoxyphenol (syringol) | 91-10-1 | MFCD00064434 | 154.16 | 1 G
SDP
Catalog No. 5000449130
Encompass_Preferred
2,6-Dimethoxyphenol, also known as Syringol, is a phenolic compound recognized for its free radical scavenging properties and antioxidant activity. It serves as a valuable substrate in assays for determining laccase activity.
  • Scavenges free radicals
  • Exhibits antioxidant activity
  • Functions as a substrate for laccase activity determination
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO and water
  • Demonstrates biological activity, inhibiting N-nitrosomorpholine formation in rat models
Catalog No. 50-004-49130 Supplier Medchemexpress LLC Supplier No. HYW0039721G
2,6-Dimethoxyphenol, also known as Syringol, is a phenolic compound recognized for its free radical scavenging properties and antioxidant activity. It serves as a valuable substrate in assays for determining laccase activity.
  • Scavenges free radicals
  • Exhibits antioxidant activity
  • Functions as a substrate for laccase activity determination
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO and water
  • Demonstrates biological activity, inhibiting N-nitrosomorpholine formation in rat models