Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxytoluene 98.0+%, TCI America™
SDP

Catalog No. D224610G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D224610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D224610G Supplier TCI America Supplier No. D224610G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5673-07-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00008374
InChI Key FPEUDBGJAVKAEE-UHFFFAOYSA-N
Synonym 2,6-dimethoxytoluene, benzene, 1,3-dimethoxy-2-methyl, 2-methyl-1,3-bis methyloxy benzene, 1,3-dimethoxy-2-methylbenzol, pubchem4545, acmc-1aqu0, 3-chloro-6-hydroxybiphenyl, 1,3-dimethoxy-2-methyl-benzene, #
PubChem CID 79755
IUPAC Name 1,3-dimethoxy-2-methylbenzene
SMILES CC1=C(C=CC=C1OC)OC

Specifications

Melting Point 39°C
Color White
Boiling Point 222°C
Quantity 10 g
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethoxytoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.