Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyl-3-heptene (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. D126201ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D126201ML 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D126201ML Supplier TCI America Supplier No. D12620.1ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2738-18-3
Molecular Formula C9H18
Molecular Weight (g/mol) 126.243
MDL Number MFCD00026523
InChI Key KDISTZUHDQPXDE-WAYWQWQTSA-N
PubChem CID 10953509
IUPAC Name (Z)-2,6-dimethylhept-3-ene
SMILES CC(C)CC=CC(C)C

Specifications

Color Yellow
Boiling Point 134°C
Quantity 0.1 mL
UN Number 3295
Formula Weight 126.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dimethyl-3-heptene (cis- and trans- mixture)
EINECSNumber (2)-0025
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.