Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,6-Dimethyl-4-heptanone

Catalog No. 5376911L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
18 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-769-11L 1 L
53-769-118L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-769-11L Supplier Sigma Aldrich Supplier No. 5376911L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,6-dimethyl-4-heptanone (di-i-butyl ketone) is widely employed as an industrial solvent
  • It undergoes gas-phase reaction with the oh radical in the presence of nox to afford acetone and 2-methylpropanal
  • 2,6-dimethyl-4-heptanone is formed as a product during the thermal degradation of yttrium(iii) isovalerate y.2,6-dimethyl-4-heptanone may be employed as a diluent for the isolation of pentanedioic acid by using amberlite la-2

Specifications

CAS 108-83-8
Density 0.808 g/mL (at 25°C (literature))
Boiling Point 165°C to 170°C (lit.)
Molecular Formula C9H18O
Refractive Index n20/D 1.412 (literature)
Linear Formula (CH3)2 CHCH2COCH2CH(CH3)2
MDL Number MFCD00008940
Quantity 1 L
Synonym Diisobutyl ketone; Isovalerone
Molecular Weight (g/mol) 142.24
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.