Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,6-Dimethyl-5-heptenal

Catalog No. 30796325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-796-325G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 30-796-325G Supplier Sigma Aldrich Supplier No. 30796325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,6-Dimethyl-5-heptenal

Specifications

CAS 106-72-9
Density 0.879 g/mL (at 25°C)
Boiling Point 116°C to 124°C (100 mmHg)
Molecular Formula C9H16O
Refractive Index n20/D 1.444 (literature)
Linear Formula (CH3)2 C=CHCH2CH2CH(CH3)CHO
MDL Number MFCD00006981
Quantity 250 mg
Synonym Melonal
Molecular Weight (g/mol) 140.22
Percent Purity 80%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.