Learn More
Medchemexpress LLC 2,6-Dimethyl-L-tyrosine (Dmt) | 123715-02-6 | 209.24 | 5 G
SDP
Catalog No. 5000446312
Encompass_Preferred
2,6-Dimethyl-L-tyrosine (Dmt) is a tyrosine derivative designed to enhance receptor affinity, functional bioactivity, and in vivo analgesia of opioid peptides. It is intended for research use only.
  • Enhances receptor affinity in opioid peptides
  • Improves functional bioactivity
  • Provides in vivo analgesia
  • Suitable for research applications
Catalog No. 50-004-46312 Supplier Medchemexpress LLC Supplier No. HYW0006655G
2,6-Dimethyl-L-tyrosine (Dmt) is a tyrosine derivative designed to enhance receptor affinity, functional bioactivity, and in vivo analgesia of opioid peptides. It is intended for research use only.
  • Enhances receptor affinity in opioid peptides
  • Improves functional bioactivity
  • Provides in vivo analgesia
  • Suitable for research applications